Anti-CYP46A1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301675EA 569 EUR
ORIGTA301675
Anti-CYP46A1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: CYP46A1 (cytochrome P450, family 46, subfamily A, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR