Imprimer

Anti-PLIN3 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA301687EA 658 EUR
ORIGTA301687
Anti-PLIN3 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: PLIN3 (perilipin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR