Anti-NPM1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA301725EA 601 EUR
ORIGTA301725
Anti-NPM1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1: 1000; Immunochemistry: 1:50-1: 100; Flow Cytometry: 1: 100

Type: Primary
Antigen: NPM1 (nucleophosmin (nucleolar phosphoprotein B23, numatrin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR