Anti-RPS6KB2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302019EA 638 EUR
ORIGTA302019
Anti-RPS6KB2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: RPS6KB2 (ribosomal protein S6 kinase, 70kDa, polypeptide 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR