Imprimer

Anti-SIGMAR1 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302032EA 698 EUR
ORIGTA302032
Anti-SIGMAR1 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SIGMAR1 (sigma non-opioid intracellular receptor 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR