Imprimer

Anti-ISG15 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302035EA 698 EUR
ORIGTA302035
Anti-ISG15 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: ISG15 (ISG15 ubiquitin-like modifier)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR