Imprimer

Anti-SNAI1 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302097EA 698 EUR
ORIGTA302097
Anti-SNAI1 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: SNAI1 (snail family zinc finger 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR