Imprimer

Anti-PIK3R1 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302158EA 698 EUR
ORIGTA302158
Anti-PIK3R1 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: PIK3R1 (phosphoinositide-3-kinase, regulatory subunit 1 (alpha))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR