Anti-MAPK14 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302203EA 638 EUR
ORIGTA302203
Anti-MAPK14 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Flow Cytometry: 1:10-50

Type: Primary
Antigen: MAPK14 (mitogen-activated protein kinase 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR