Imprimer

Anti-NCOA1 Rabbit Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302235EA 698 EUR
ORIGTA302235
Anti-NCOA1 Rabbit Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: NCOA1 (nuclear receptor coactivator 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR