Anti-PIK3CG Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA302283EA 638 EUR
ORIGTA302283
Anti-PIK3CG Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: PIK3CG (phosphatidylinositol-4,5-bisphosphate 3-kinase, catalytic subunit gamma)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR