Imprimer

Anti-CARD11 Goat polyclonal antibody

Fournisseur: OriGene
ORIGTA302440EA 547 EUR
ORIGTA302440
Anti-CARD11 Goat polyclonal antibody
Anticorps
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: CARD11 (caspase recruitment domain family, member 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR