Imprimer

Anti-CHRM1 Goat polyclonal antibody

Fournisseur: OriGene
ORIGTA302450EA 547 EUR
ORIGTA302450
Anti-CHRM1 Goat polyclonal antibody
Anticorps
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: CHRM1 (cholinergic receptor, muscarinic 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR