Anti-ALOX5AP Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302488EA 502 EUR
ORIGTA302488
Anti-ALOX5AP Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: ALOX5AP (arachidonate 5-lipoxygenase-activating protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR