Anti-IDE Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302516EA 502 EUR
ORIGTA302516
Anti-IDE Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: IDE (insulin-degrading enzyme)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR