Imprimer

Anti-KPNA4 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302525EA 547 EUR
ORIGTA302525
Anti-KPNA4 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1/32000 Western Blot: 02-1µg/ml; Immunochemistry: 3µg/ml

Type: Primary
Antigen: KPNA4 (karyopherin alpha 4 (importin alpha 3))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR