Anti-SH2B3 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302531EA 500 EUR
ORIGTA302531
Anti-SH2B3 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-10 µg/ml

Type: Primary
Antigen: SH2B3 (SH2B adaptor protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR