Anti-PPARD Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302579EA 500 EUR
ORIGTA302579
Anti-PPARD Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: PPARD (peroxisome proliferator-activated receptor delta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR