Anti-RBP4 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302591EA 500 EUR
ORIGTA302591
Anti-RBP4 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:2000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: RBP4 (retinol binding protein 4, plasma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR