Anti-HTR3A Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302602EA 500 EUR
ORIGTA302602
Anti-HTR3A Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: HTR3A (5-hydroxytryptamine (serotonin) receptor 3A, ionotropic)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR