Anti-SLC9A3R2 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302611EA 500 EUR
ORIGTA302611
Anti-SLC9A3R2 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: SLC9A3R2 (solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 regulator 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR