Imprimer

Anti-TRAF2 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302626EA 547 EUR
ORIGTA302626
Anti-TRAF2 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:128000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: TRAF2 (TNF receptor-associated factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR