Imprimer

Anti-GC Goat polyclonal antibody

Fournisseur: OriGene
ORIGTA302630EA 547 EUR
ORIGTA302630
Anti-GC Goat polyclonal antibody
Anticorps
Recommended Dilutions: ELISA: 1:2000 Western Blot: 01-03µg/ml

Type: Primary
Antigen: GC (group-specific component (vitamin D binding protein))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR