Anti-CFL2 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302652EA 502 EUR
ORIGTA302652
Anti-CFL2 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000; Western Blot: 001-003µg/ml

Type: Primary
Antigen: CFL2 (cofilin 2 (muscle))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR