Anti-MAPK3 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302659EA 438 EUR
ORIGTA302659
Anti-MAPK3 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:4000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: MAPK3 (mitogen-activated protein kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR