Imprimer

Anti-IRAK4 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302664EA 547 EUR
ORIGTA302664
Anti-IRAK4 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: IRAK4 (interleukin-1 receptor-associated kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR