Anti-GRB2 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302665EA 438 EUR
ORIGTA302665
Anti-GRB2 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 005-015?g/ml; Immunochemistry: 01-03?g/ml

Type: Primary
Antigen: GRB2 (growth factor receptor-bound protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR