Anti-IRF5 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302667EA 438 EUR
ORIGTA302667
Anti-IRF5 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 01-03?g/ml

Type: Primary
Antigen: IRF5 (interferon regulatory factor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR