Anti-XRCC6 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302668EA 438 EUR InStock
ORIGTA302668
Anti-XRCC6 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:2000; Western Blot: 01-03µg/ml

Type: Primary
Antigen: XRCC6 (X-ray repair complementing defective repair in Chinese hamster cells 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR