Anti-JUP Goat polyclonal antibody

Supplier: OriGene
ORIGTA302670EA 438 EUR
ORIGTA302670
Anti-JUP Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 02-1µg/ml

Type: Primary
Antigen: JUP (junction plakoglobin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR