Imprimer

Anti-LEPR Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302673EA 547 EUR
ORIGTA302673
Anti-LEPR Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: LEPR (leptin receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR