Anti-BIRC7 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302674EA 500 EUR
ORIGTA302674
Anti-BIRC7 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: BIRC7 (baculoviral IAP repeat containing 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR