Imprimer

Anti-RANBP1 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302686EA 550 EUR
ORIGTA302686
Anti-RANBP1 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 0.05-0.1?g/ml; Immunochemistry: 3-5?g/ml; ELISA: 1:16000

Type: Primary
Antigen: RANBP1 (RAN binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR