Anti-RANGAP1 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302687EA 500 EUR
ORIGTA302687
Anti-RANGAP1 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 01-05µg/ml

Type: Primary
Antigen: RANGAP1 (Ran GTPase activating protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR