Anti-UBE2L3 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302697EA 438 EUR InStock
ORIGTA302697
Anti-UBE2L3 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000; Western Blot: 05-2µg/ml

Type: Primary
Antigen: UBE2L3 (ubiquitin-conjugating enzyme E2L 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR