Anti-VDAC2 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302698EA 438 EUR
ORIGTA302698
Anti-VDAC2 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:64000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: VDAC2 (voltage-dependent anion channel 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Dog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR