Imprimer

Anti-WNT4 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302700EA 547 EUR
ORIGTA302700
Anti-WNT4 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 1-3.0?g/ml

Type: Primary
Antigen: WNT4 (wingless-type MMTV integration site family, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR