Imprimer
Anti-PGAM1 / PGAM2 / PGAM4 Goat Polyclonal Antibody
Fournisseur: OriGene
ORIGTA302705EA 547 EUR
ORIGTA302705
Anti-PGAM1 / PGAM2 / PGAM4 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:32000; Western Blot: 05-2µg/ml
Type: Primary
Antigen: PGAM1 / PGAM2 / PGAM4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human