Anti-DDB1 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302707EA 438 EUR
ORIGTA302707
Anti-DDB1 Goat polyclonal antibody
Antibodies
Recommended Dilutions: Western Blot: 02-1 µg/ml

Type: Primary
Antigen: DDB1 (damage-specific DNA binding protein 1, 127kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR