Anti-FEM1C Goat polyclonal antibody

Supplier: OriGene
ORIGTA302709EA 438 EUR
ORIGTA302709
Anti-FEM1C Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:8000; Western Blot: 03-1µg/ml

Type: Primary
Antigen: FEM1C (fem-1 homolog c (C. elegans))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR