Anti-ORC3 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302714EA 438 EUR
ORIGTA302714
Anti-ORC3 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: ORC3 (origin recognition complex, subunit 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR