Anti-STAT3 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302716EA 438 EUR InStock
ORIGTA302716
Anti-STAT3 Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:128000; Western Blot: 1-3µg/ml; Immunochemistry: 10?g/ml

Type: Primary
Antigen: STAT3 (signal transducer and activator of transcription 3 (acute-phase response factor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR