Imprimer

Anti-CAPZB Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302723EA 547 EUR
ORIGTA302723
Anti-CAPZB Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1/32000; Western Blot: 05-2µg/ml

Type: Primary
Antigen: CAPZB (capping protein (actin filament) muscle Z-line, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR