Anti-SMAD4 Goat Polyclonal Antibody

Supplier: OriGene
ORIGTA302724EA 500 EUR
ORIGTA302724
Anti-SMAD4 Goat Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 05-2µg/ml

Type: Primary
Antigen: SMAD4 (SMAD family member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR