Anti-FYN Goat polyclonal antibody

Supplier: OriGene
ORIGTA302725EA 438 EUR
ORIGTA302725
Anti-FYN Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:32000; Western Blot: 02-1µg/ml

Type: Primary
Antigen: FYN (FYN oncogene related to SRC, FGR, YES)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR