Imprimer

Anti-DOCK1 Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302729EA 547 EUR
ORIGTA302729
Anti-DOCK1 Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: Western Blot: 3-4µg/ml

Type: Primary
Antigen: DOCK1 (dedicator of cytokinesis 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR