Anti-TIRAP Goat polyclonal antibody

Supplier: OriGene
ORIGTA302730EA 438 EUR
ORIGTA302730
Anti-TIRAP Goat polyclonal antibody
Antibodies
Recommended Dilutions: ELISA: 1:16000 Western Blot: 1-3µg/ml

Type: Primary
Antigen: TIRAP (toll-interleukin 1 receptor (TIR) domain containing adaptor protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR