Anti-NFIL3 Goat polyclonal antibody

Supplier: OriGene
ORIGTA302736EA 438 EUR
ORIGTA302736
Anti-NFIL3 Goat polyclonal antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3µg/ml

Type: Primary
Antigen: NFIL3 (nuclear factor, interleukin 3 regulated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR