Imprimer

Anti-PPP2CA / PPP2CB Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302738EA 547 EUR
ORIGTA302738
Anti-PPP2CA / PPP2CB Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:128000; Western Blot: 1-3µg/ml

Type: Primary
Antigen: PPP2CA / PPP2CB
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR