Imprimer

Anti-PPP2R5A Goat Polyclonal Antibody

Fournisseur: OriGene
ORIGTA302741EA 547 EUR
ORIGTA302741
Anti-PPP2R5A Goat Polyclonal Antibody
Anticorps
Recommended Dilutions: ELISA: 1:32000; Western Blot: 05-2µg/ml

Type: Primary
Antigen: PPP2R5A (protein phosphatase 2, regulatory subunit B', alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR